Order Entry
Belgium
ContactUsLinkComponent
Analytes C for ICP
Analytes C for ICP
Catalog # HONCE2120-100ML
CAS Number:  
Analytes C for ICP
Catalog # HONCE2120-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Analytes C

Specifications

About this item

Certified Reference Material (CRM).

2 mg/L each of Ag, As, Ba, Be, Cd, Co, Cr, Cu, Hg, Mn, Ni, Pb, Sb, Se, Tl, V, Zn in HNO₃ 2%