Order Entry
Belgium
ContactUsLinkComponent
Alternate Analytes B for ICP
Alternate Analytes B for ICP
Catalog # HONCE2117-100ML
CAS Number:  
Alternate Analytes B for ICP
Catalog # HONCE2117-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Alternate Analytes B

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Al, As, B, Mo, Na, Sb, Se, Tl, 10 mg/L each of Ca, Fe, Mg, Si in HNO₃ 5%