Order Entry
Belgium
ContactUsLinkComponent
Analytes B for ICP
Analytes B for ICP
Catalog # HONCE2116-100ML
CAS Number:  
Analytes B for ICP
Catalog # HONCE2116-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Analytes B

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Cd, Ni, Zn, 60 mg/L Sb, 50 mg/L each of Ba, Be, Co, Cr, Cu, Mn, V, 20 mg/L Ag, 10 mg/L each of As, Tl, 5 mg/L each of Pb, Se in HNO₃ 5%