Order Entry
Belgium
ContactUsLinkComponent
Alternate Interference A for ICP
Alternate Interference A for ICP
Catalog # HONCE2115-100ML
CAS Number:  
Alternate Interference A for ICP
Catalog # HONCE2115-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Alternate Interference A

Specifications

About this item

Certified Reference Material (CRM).

1000 mg/L each of Cr, Cu, Mn, Ni, Ti, V in HNO₃ 5%