ContactUsLinkComponent
Primary Interferents Interferents A for ICP
Primary Interferents Interferents A for ICP
  HONCE2114-100ML
 :  
Primary Interferents Interferents A for ICP
  HONCE2114-100ML
 :  
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Primary Interferents Interferents A
Certified Reference Material (CRM).
5000 mg/L each of Al, Ca, Mg, 2000 mg/L Fe in HNO₃ 5%