Order Entry
Belgium
ContactUsLinkComponent
Interference check standard 18 for ICP
Interference check standard 18 for ICP
Catalog # HONCE2113-100ML
CAS Number:  
Interference check standard 18 for ICP
Catalog # HONCE2113-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Interference Check Standard 18

Specifications

About this item

Certified Reference Material (CRM).

20000 mg/L K, 1000 mg/L each of As, Pb, Tl, 500 mg/L Se, 300 mg/L each of Ag, Ba, Cd, Co, Cr, Cu, Ni, V, Zn, 200 mg/L Mn, 100 mg/L each of Be, Hg in HNO₃ 5%