Order Entry
Belgium
ContactUsLinkComponent
EPA 6010 interference check standard for ICP-AES
EPA 6010 interference check standard for ICP-AES
Catalog # HONCE2112-100ML
CAS Number:  
EPA 6010 interference check standard for ICP-AES
Catalog # HONCE2112-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    EPA 6010 Interference Check Standard

Specifications

About this item

Certified Reference Material (CRM).

6000 mg/L Ca, 5000 mg/L Fe, 3000 mg/L Mg, 1200 mg/L Al, 1000 mg/L Na in HNO₃ 5%