Order Entry
Belgium
ContactUsLinkComponent
Contract required detection limits (CRDL) ICP and HGA elements
Contract required detection limits (CRDL) ICP and HGA elements
Catalog # HONCE2107-100ML
CAS Number:  
Contract required detection limits (CRDL) ICP and HGA elements
Catalog # HONCE2107-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Contract Required Detection Limits (CRDL) ICP and HGA elements

Specifications

About this item

Certified Reference Material (CRM).

120 mg/L Sb, 100 mg/L each of Co, V, 80 mg/L Ni, 50 mg/L Cu, 40 mg/L Zn, 30 mg/L Mn, 20 mg/L each of Ag, As, Cr, Tl, 10 mg/L each of Be, Cd, Se, 6 mg/L Pb in HNO₃ 5%