ContactUsLinkComponent
Instrument Calibration standard 2 for ICP and HGA elements
  HONCE2104-100ML
 :  
Instrument Calibration standard 2 for ICP and HGA elements
  HONCE2104-100ML
 :  
  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Instrument Calibration Standard 2
Certified Reference Material (CRM).
400 mg/L Ni, 200 mg/L Zn, 150 mg/L Mn, 100 mg/L each of Ag, Cr in HNO₃ 5%