Order Entry
Belgium
ContactUsLinkComponent
Quality control standard 7A elements for ICP
Quality control standard 7A elements for ICP
Catalog # HONCE2102-100ML
CAS Number:  
Quality control standard 7A elements for ICP
Catalog # HONCE2102-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Quality Control Standard 7A Elements

Specifications

About this item

Certified Reference Material (CRM).

1000 mg/L K, 500 mg/L Si, 100 mg/L each of Al, B, Ba, Na, 50 mg/L Ag in HNO₃ 5%