Order Entry
Belgium
Orders LinkContactUsLinkComponent
UV Wavecal solution for ICP

UV Wavecal solution for ICP

UV Wavecal Solution, 2% HCl, Plastic bottle, 100 mL
Catalog # HONCE2099-100ML
Supplier:   Honeywell Chemicals
UV Wavecal solution for ICP
Catalog # HONCE2099-100ML
CAS Number:  
UV Wavecal solution for ICP

UV Wavecal Solution, 2% HCl, Plastic bottle, 100 mL

CAS Number:  
Price
226,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

100 mg/L each of K, P, S, 20 mg/L each of As, La, Li, Mn, Mo, Na, Ni, Sc in HCl 2%

Specifications

Specifications

  • Matrix:
    2% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    UV Wavecal Solution

Specifications