Order Entry
Belgium
ContactUsLinkComponent
Mixed calibration standard 3 for ICP
Mixed calibration standard 3 for ICP
Catalog # HONCE2096-100ML
CAS Number:  
Mixed calibration standard 3 for ICP
Catalog # HONCE2096-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Mixed Calibration Standard 3

Specifications

About this item

Certified Reference Material (CRM).

500mg/L As, 100mg/L Mo, 100mg/L Si in HNO₃ 2%