Order Entry
Belgium
ContactUsLinkComponent
Mixed calibration standard for ICP
Mixed calibration standard for ICP
Catalog # HONCE2093-100ML
CAS Number:  
Mixed calibration standard for ICP
Catalog # HONCE2093-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Mixed Calibration Standard

Specifications

About this item

Certified Reference Material (CRM).

50 mg/L each of As, K, 10 mg/L each of La, Li, Mn, Ni, Sr, Zn, 1 mg/L each of Ba, Mg in HNO₃ 2%