Combined Six Cation Standard II for IC
HONCE2033-100ML
:
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Combined Six Cation Standard II
Certified Reference Material (CRM).
500 mg/L each of K+, Ca2+, 250 mg/L each of NH4+, Mg2+, 200 mg/L Na+, 50 mg/L Li+ in H₂O