Order Entry
Belgium
ContactUsLinkComponent
Multi-element calibration standard 3 for ICP
Multi-element calibration standard 3 for ICP
Catalog # HONCE2027-100ML
CAS Number:  
Multi-element calibration standard 3 for ICP
Catalog # HONCE2027-100ML
CAS Number:  

Specifications

  • Matrix:
    10% HCl/1% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Calibration Standard 3

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn in HCl 10% / HNO₃ 1%