Order Entry
Belgium
ContactUsLinkComponent
Calibration mix 2 for AA and ICP-OES
Calibration mix 2 for AA and ICP-OES
Catalog # HONCE2021-100ML
CAS Number:  
Calibration mix 2 for AA and ICP-OES
Catalog # HONCE2021-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Calibration Mix 2

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Ni, Pb, Se, Th, Tl, U, V, Zn in HNO₃ 5%