Order Entry
Belgium
ContactUsLinkComponent
ICP-OES Wavelength calibration Concentrate
ICP-OES Wavelength calibration Concentrate
Catalog # HONCE2015-100ML
CAS Number:  
ICP-OES Wavelength calibration Concentrate
Catalog # HONCE2015-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    ICP-OES Wavelength Calibration Concentrate

Specifications

About this item

Certified Reference Material (CRM).

500mg/L K, 50 mg/L each of Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn in HNO₃ 5%