Order Entry
Belgium
ContactUsLinkComponent
ICP-MS stock tuning solution
ICP-MS stock tuning solution
Catalog # HONCE2008-100ML
CAS Number:  
ICP-MS stock tuning solution
Catalog # HONCE2008-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    ICP-MS Stock Tuning Solution

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Li, Y, Ce, Co, Tl in HNO₃ 2%