Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

ICP-MS tuning solution

ICP-MS Tuning Solution, 2% HNO₃, Plastic bottle, 100 mL
Catalog # HONCE2007-100ML
Supplier:   Honeywell Chemicals
ICP-MS tuning solution
Catalog # HONCE2007-100ML
CAS Number:  
ICP-MS tuning solution

ICP-MS Tuning Solution, 2% HNO₃, Plastic bottle, 100 mL

CAS Number:  
Price
391,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Material (CRM).

10 mg/L each of Li , Mg, Y, Ce, Tl, Co in HNO₃ 2%

Specifications

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    ICP-MS Tuning Solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.