Order Entry
Belgium
ContactUsLinkComponent
Initial calibration verification standard for ICP-MS
Initial calibration verification standard for ICP-MS
Catalog # HONCE2003-100ML
CAS Number:  
Initial calibration verification standard for ICP-MS
Catalog # HONCE2003-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Initial Calibration Verification Standard

Specifications

About this item

Certified Reference Material (CRM).

1000 mg/L each of Ca, Fe, K, Mg, Na, Sr, 10 mg/L each of Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Tl, V, Zn, Th, U in HNO₃ 5%