Order Entry
Belgium
ContactUsLinkComponent
Multi-element calibration standard 2A for ICP-MS
Multi-element calibration standard 2A for ICP-MS
Catalog # HONCE2001-100ML
CAS Number:  
Multi-element calibration standard 2A for ICP-MS
Catalog # HONCE2001-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Calibration Standard 2A

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, Hg, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn in HNO₃ 5%