ContactUsLinkComponent
Pocket refractormeter, IR Brix meter (Strawberry), PAL-Hikari 4
Pocket refractormeter, IR Brix meter (Strawberry), PAL-Hikari 4
Catalog # ATAG5454
Supplier:  Atago
Pocket refractormeter, IR Brix meter (Strawberry), PAL-Hikari 4
Catalog # ATAG5454
Supplier:  Atago
Specifications
  • Description:
    Pocket IR Brix Meter PAL-Hikari 4 (Strawberry)
  • Pk:
    1
Specifications
About this item
Non-destructive strawberry Brix meter.
  • The Brix (sugar level) can be measured by placing the fruit on the sample stage with no need of cutting or squeezing
It requires no more cumbersome wiping and cleansing after each measurement.
Ordering information: Requires 3 ˣ AAA batteries.