Buffer solutions, colour coded, certified (20 °C), Fluka™
Frequently Bought Together
pH buffer solutions
: Mettler - Toledo
pH buffers, ready to use
: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ
Sodium methoxide 30% (w/w) 5.4 M in methanol, AcroSeal®
: Thermo Fisher Scientific