Order Entry
Belgium
ContactUsLinkComponent
Palladium(II) nitrate 5 g/L in nitric acid 1% matrix modifier solution, for atomic absorption spectroscopy, Fluka™
Palladium(II) nitrate 5 g/L in nitric acid 1% matrix modifier solution, for atomic absorption spectroscopy, Fluka™
Catalog # HONCE4603-50ML
CAS Number:  
Palladium(II) nitrate 5 g/L in nitric acid 1% matrix modifier solution, for atomic absorption spectroscopy, Fluka™
Catalog # HONCE4603-50ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Matrix:
    HNO₃ 1%
  • Concentration:
    Pd(NO₃)₂ 5000 mg/L
  • Quality:
    ISO/IEC17025:2017(ANAB),ISO17034:2016(ANAB)
  • Form:
    Solution
  • Density:
    1.01g/cm3

Specification Test Results

About this item

 :
 :230.43 g/mol
Density:1.01 g/cm3
Storage Temperature:Ambient
CAS Number:10102-05-3
Formula:N₂O₆Pd
MDL Number:MFCD00011169