Order Entry
Belgium
ContactUsLinkComponent
 
Magnesium nitrate 10 g/L in water matrix modifier solution, for atomic absorption spectroscopy, Fluka™
  HONCE4600-50ML
 :  
Magnesium nitrate 10 g/L in water matrix modifier solution, for atomic absorption spectroscopy, Fluka™
  HONCE4600-50ML
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

 

 

  • Matrix:
    H₂O
  • Concentration:
    Mg(NO₃)₂ 10 g/L
  • Quality:
    ISO/IEC17025:2017(ANAB),ISO17034:2016(ANAB)
  • Form:
    Solution
  • Density:
    1.01g/cm3

 

 

 :
 :1.01 g/cm3
 :Ambient
 :13446-18-9
 :Mg(NO₃)₂