Order Entry
Belgium
Orders LinkContactUsLinkComponent
HyClone Cell Boost™ 3 supplement

HyClone Cell Boost™ 3 supplement

HyClone Cell Boost™ 3 supplement
Certificates
HyClone Cell Boost™ 3 supplement

2 Options Available Below

Product Details & Documents

Supplements cell culture with amino acids, vitamins, glucose, and trace elements and manufactured to meet cGMP manufacturing standards and QC specifications.

  • Designed to support modern manufacturing processes, Cell Boost 3 is chemically defined (CD), animal-derived component-free (ADCF), protein-free, and hydrolysate-free
  • For convenience, this cell culture supplement is available as a ready-to-use liquid solution in addition to a dry powder format

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 3 Supplement provides amino acids, vitamins, glucose, and trace elements to support fed-batch cultures in research and bioprocessing applications. It is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

This HyClone cell culture supplement has been developed for recombinant protein production with various cell lines including hybridoma and myeloma cell lines.

Supplement 20 to 200 ml/L one or more times, starting during mid to late growth phase. This is expected to be day 3 to 4. Start the culture at a reduced volume according to anticipated feed volumes. Continue supplementation daily until the final volume is reached. It is recommended to monitor L- glutamine separately and supplement as needed.

Dry powdered medium milling equivalency study compares pin milling of powdered HyClone cell culture media and supplements at two manufacturing sites. Detailed physical, chemical, and biological data, including cell growth, are provided.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SH31115.02
  • HYCLSH30825.01
  • Storage temperature:
  • 2…8 °C
  • 2…8 °C

Specifications

Product Family Options

Product Information

  • Form
    Size
    Pk
    Availability
    Price
  • SH31115.02
    Cell Boost™ 3
    Cell Boost™ 3
    Liquid
    1000 ml
    1.000 mL
    Log in for availability
    Log in for availability
    1 * 1.000 mL
    1 * 1.000 mL - 183,00 €
    183,00 €
    1 * 1.000 mL
       183,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SH31115.02
    Storage temperature
    2…8 °C
  • HYCLSH30825.01
    Cell Boost™ 3
    Cell Boost™ 3
    Powder
    100 g
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 219,00 €
    219,00 €
    1 * 100 g
       219,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    HYCLSH30825.01
    Storage temperature
    2…8 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.