ContactUsLinkComponent
Microscopes, Panthera TEC POL series
Microscopes, Panthera TEC POL series
Supplier:  MOTIC
Certificates
undefined
Microscopes, Panthera TEC POL series
Supplier:  MOTIC
Specifications
  • Catalog No:
  • 630-3120
  • 630-3119
  • 630-3121
  • 630-3118
  • 630-3124
  • Model:
  • Panthera TEC POL Epi Binocular
  • Panthera TEC POL Trinocular
  • Panthera TEC POL Epi Trinocular
  • Panthera TEC POL Binocular
  • Panthera TEC POL (i) Digital with integrated WiFi camera
  • Observation method:
  • Brightfield and polarisation
  • Brightfield and polarisation
  • Brightfield and polarisation
  • Brightfield and polarisation
  • Brightfield and polarisation
  • Head:
  • 25° inclined, 360° rotating
  • 25° inclined, 360° rotating
  • 25° inclined, 360° rotating
  • 25° inclined, 360° rotating
  • 25° inclined, 360° rotating
  • Eyepieces:
  • UC-WF 10×/22 mm
  • UC-WF 10×/22 mm
  • UC-WF 10×/22 mm
  • UC-WF 10×/22 mm
  • UC-WF 10×/22 mm
  • Nosepiece:
  • Quintuple, reversed
  • Quintuple, reversed
  • Quintuple, reversed
  • Quintuple, reversed
  • Quintuple, reversed
  • Objectives:
  • Achromatic 4×, 10×, 40×, 60×
  • Achromatic 4×, 10×, 40×, 60×
  • Achromatic 4×, 10×, 40×, 60×
  • Achromatic 4×, 10×, 40×, 60×
  • Achromatic 4×, 10×, 40×, 60×
  • Stage:
  • Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
  • Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
  • Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
  • Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
  • Circular rotating 360°, lockable, Ø 160 mm, 1° increments, 0,1° vernier scale
  • Focusing system:
  • Coaxial coarse and fine focusing system with tension adjustment
  • Coaxial coarse and fine focusing system with tension adjustment
  • Coaxial coarse and fine focusing system with tension adjustment
  • Coaxial coarse and fine focusing system with tension adjustment
  • Coaxial coarse and fine focusing system with tension adjustment
  • Camera:
  • None
  • None
  • None
  • None
  • Integrated camera with chip ¹/₃", data transfer: Wi-Fi, ethernet. Capture resolution 4 MP (2592×1520), live display mode via WiFi or ethernet 1280×720, 1920×1080 (full HD)
  • Weight:
  • 15,4 kg
  • 15,4 kg
  • 15,4 kg
  • 15,4 kg
  • 15,4 kg
Specifications
Eyepieces
Product Information
Frequently Bought Together