Order Entry
Belgium
ContactUsLinkComponent
 
 
Trisodium phosphate dodecahydrate 98.0-102.0%, AnalaR NORMAPUR® Reag. Ph.Eur., ACS
 
undefined
 
Trisodium phosphate dodecahydrate 98.0-102.0%, AnalaR NORMAPUR® Reag. Ph.Eur., ACS
 

 

  • Assay                            :
    98.0 - 102.0 %
  • Excess alkali (as NaOH) :
    Max. 2.5 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insoluble matter:
    Max. 100 ppm
  • Chloride (Cl):
    Max. 10 ppm
  • Sulphate (SO₄):
    Max. 100 ppm
  • Iron (Fe)   :
    Max. 10 ppm
  • Conforms to Reag. Ph.Eur./ACS :
    Passes test

 

Frequently Bought Together