Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Soy bean Lipoxidase
Soy bean Lipoxidase
Soy bean Lipoxidase
Catalog # TCIAL0059-100MG
Supplier:
TCI
CAS Number:
Expand image
Soy bean Lipoxidase
Catalog # TCIAL0059-100MG
Supplier:
TCI
CAS Number:
Specifications
Pk:
100 mg
Enzyme name:
Lipoxidase
Specifications
About this item
Product Details
Product Details