ContactUsLinkComponent
Avantor® Hichrom HI-DEX DAC Gamma, GC Columns
 :  Avantor
undefined
Avantor® Hichrom HI-DEX DAC Gamma, GC Columns
 :  Avantor
  • Catalog No:
  • HI67-25-025-15
  • HI67-25-025-25
  • HI67-25-015-60
  • HI67-25-025-60
  • HI79-10-010-10
  • HI67-25-025-30
  • HI67-25-025-50
  • HI67-25-015-30
  • HI67-25-015-50
  • HI67-25-015-15
  • HI79-10-010-5
  • HI67-25-015-25
  • Phase name:
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • HI-DEX DAC Gamma
  • Phase composition:
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Diacethyl TBS Gamma Cyclodexstrin based
  • Polarity:
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Chiral stationary phase
  • Crossbond:
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Temperature limit:
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
  • 230 °C
Frequently Bought Together