Orders LinkContactUsLinkComponent
Spill workfloor
Spill workfloor
Spill workfloor heavy for 4 drums, 1600×1600×150 mm
Catalog # BRDY196262
Supplier:   Brady
Spill workfloor
Catalog # BRDY196262
Supplier:  Brady
Spill workfloor
Spill workfloor
Spill workfloor heavy for 4 drums, 1600×1600×150 mm
Price
733,74 €
Log in for availability
1 * 1 ST
Product Details & Documents
Workfloors are a great way to handle and store hazardous fluids and contain potentially hazardous leaks in storage and dispensing areas.
  • Can hold/carry drums or smaller containers
  • Can be easily assembled with other units by using the connector system
  • Compatible with most forklifts and pallet trucks
Specifications
  • Description:
    Spill workfloor heavy for 4 drums
  • W×D×H:
    1600×1600×150 mm