ContactUsLinkComponent
Spill berm, QuickBerm® rigid lock, EG-5
Spill berm, QuickBerm® rigid lock, EG-5
Catalog # BRDY198532
Supplier:  Brady
Spill berm, QuickBerm® rigid lock, EG-5
Catalog # BRDY198532
Supplier:  Brady
Specifications
  • Capacity:
    890 L
  • Description:
    Spill berm, QuickBerm® rigid lock, EG-5
  • W×D×H:
    1422×2642×305 mm
  • Pk:
    1
Specifications
About this item
The extreme durable Rigid-Lock QuickBerm® provided immediate, secure deployment and spill-readiness. The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.
  • Made of PVC coated fabric
  • Black in colour
The high performance material withstands a broad range of chemicals including petroleum, oils, grease, and most acids and a broad UV resistance - durable enough for extended wear.