Label printer, BSP61
About this item
The BSP™61 Print and apply system offers a fully automatic labelling solution anywhere along the production line. It combines a thermal transfer label printer with an automatic applicator and is designed to consistently print and accurately position and apply labels in almost all applications.
- Print speed is 250 mm/s
- Internal memory capacity is 64 mb with RAM/flash of 8 mb
- Monocolour
- Designed to handle up to 7000 labels per day
It remove the problems of automating label application to ensure reliable tracking and tracing with no cutting into the liner, clean pick up, no adhesive bleeding, no label curling and accuracy.
Higher volume applications such as PCB marking, component and product marking in high quality demanding industries like automotive, electronics and manufacturing.
Label dimension limits: Maximum width of 114 mm, minimum width of 10 mm, minimum height of 8 mm.
Ordering information: For more information on available applicators or other accessories; contact your Avantor representative.
2 Options Available Below
Product Details & Documents
The BSP™61 Print and apply system offers a fully automatic labelling solution anywhere along the production line. It combines a thermal transfer label printer with an automatic applicator and is designed to consistently print and accurately position and apply labels in almost all applications.
- Print speed is 250 mm/s
- Internal memory capacity is 64 mb with RAM/flash of 8 mb
- Monocolour
- Designed to handle up to 7000 labels per day
It remove the problems of automating label application to ensure reliable tracking and tracing with no cutting into the liner, clean pick up, no adhesive bleeding, no label curling and accuracy.
Higher volume applications such as PCB marking, component and product marking in high quality demanding industries like automotive, electronics and manufacturing.
Label dimension limits: Maximum width of 114 mm, minimum width of 10 mm, minimum height of 8 mm.
Ordering information: For more information on available applicators or other accessories; contact your Avantor representative.
Product Family Options
Product Information
- Print widthDisponibilitéPrix
- BRDY710949Label BSP61 for up to 101 mm wide consumables, 300 dpi, to be combined with left applicatorLabel BSP61 for up to 101 mm wide consumables, 300 dpi, to be combined with left applicator105,60 mmLog in for availability1 * 1 ST4 140,00 €
Applicator
Product Information
- Print widthDisponibilitéPrix
- BRDY711040Left applicator, cyl. stroke 200 mm with pressure reducerLeft applicator, cyl. stroke 200 mm with pressure reducerLog in for availability1 * 1 ST3 960,00 €
Accessories
Product Information
- N° de catalogueForDisponibilitéPrix
- Brady workstation scan and print software suiteBrady workstation scan and print software suiteBRDY149445BBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200Log in for availability1 * 1 ST - 833,00 €
Product Information
- N° de catalogueDisponibilitéPrix
- Brady workstation CLP/GHS app via download
Product Information
- N° de catalogueForDisponibilitéPrix
- Workstation lockout writer software appWorkstation lockout writer software appBRDY145445BBP®31, BBP®33, BBP®85, i3300Log in for availability1 * 1 ST - 833,00 €
Product Information
- N° de catalogueForDisponibilitéPrix
- Brady workstation automation software suiteBrady workstation automation software suiteBRDY149187BBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500Log in for availability1 * 1 ST - 833,00 €