

1-Decanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:112-30-1
:
Formula:H₃C(CH₂)₈CH₂OH
MDL Number:MFCD00004747
:158.28 g/mol
Merck Index:13,02875
Boiling Pt:230 °C (1013 hPa)
Melting Pt:7 °C
Density:0.82634 g/cm³ (20 °C)
Flash Pt:82 °C
Storage Temperature:Ambient
EINECS:203-956-9
UN:3082
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Assay:Min. 99 %
- Density (20/4):0.829 --> 0.830
Specification Test Results
Related Information
Frequently Bought Together