The BradyPrinter i7100 industrial thermal transfer bench top printer is ideal for customers requiring reliable high volume printing on lots of different materials within the aerospace, defence and mass transit, electronics, automotive, logistics, electrical, voice-datacom and lab markets.
- 300 or 600 dpi
- Applications include cable and wire identification, circuit board and component identification, and general and industrial identification
- Mono colour
It guarantees accurate printing on sleeves, tags, labels, tapes, cable markers and much more. The printer comes with the Brady Workstation Product & Wire Identification software suite enabling you to immediately start designing professional labels. This suite combines frequently used software apps to create product and cable identification labels.
Barcode types include Code 39; Code 93; Code 39 Full ASCII; code 128 A; code 128 B; code 128 C; EAN 8; EAN13; EAN/UCC 128/GS1-128; EAN/UPC; FIM; HIBC; Interleaved 2/5; ident and lead code of Deutsche Post AG; Codabar; JAN8; JAN 13; MSI; Plessey; Postnet; RSS14; UPC A; UPC E; UPC E0; Aztec; Codablock F; DataMatrix; PDF417; Micro PDF417; UPS MaxiCode; QR code; RSS 14 truncated, limited stacked and stacked omnidirectional; EAN/GS1 DataMatrix; GS1 DataBar.
Languages: Arabic (Egypt); Bulgarian; Croatian; Czech; Danish; Dutch; English; Finnish; French; German; Greek; Hungarian; Italian; Lithuanian; Macedonian; Norwegian (Bokmål); Persian (Farsi); Polish; Portuguese; Romanian; Russian; Serbian; Slovenian; Spanish; Swedish and Turkish.
- Catalog No:
- 568-0901
- 568-0895
- 568-0899
- 568-0897
- Display:
- Colour touchscreen, 272×480 pixels
- Colour touchscreen, 272×480 pixels
- Colour touchscreen, 272×480 pixels
- Colour touchscreen, 272×480 pixels
- Interfaces:
- Ethernet; I/O connection; Bluetooth (optional); WiFi (optional); Serial; USB
- Ethernet; I/O connection; Bluetooth (optional); WiFi (optional); Serial; USB
- Ethernet; I/O connection; Bluetooth (optional); WiFi (optional); Serial; USB
- Ethernet; I/O connection; Bluetooth (optional); WiFi (optional); Serial; USB
- Label size:
- Height 4 - 6 mm (for the peel-version)
- Height 4 - 6 mm (for the peel-version)
- Height 4 - 6 mm (for the peel-version)
- Height 4 - 6 mm (for the peel-version)
- Memory:
- 263 MB RAM, 50 MB IFFS Flash, SD card, slot up to 512 GB
- 256 MB RAM, 50 MB IFFS Flash, SD card, slot up to 512 GB
- 262 MB RAM, 50 MB IFFS Flash, SD card, slot up to 512 GB
- 257 MB RAM, 50 MB IFFS Flash, SD card, slot up to 512 GB
- Power supply:
- AC Adaptor, Lithium-Manganese Battery
- AC Adaptor, Lithium-Manganese Battery
- AC Adaptor, Lithium-Manganese Battery
- AC Adaptor, Lithium-Manganese Battery
- Print method:
- Thermal Transfer
- Thermal Transfer
- Thermal Transfer
- Thermal Transfer
- Software:
- Compatible with Brady Workstation, Windows based driver for 3rd party software use
- Compatible with Brady Workstation, Windows based driver for 3rd party software use
- Compatible with Brady Workstation, Windows based driver for 3rd party software use
- Compatible with Brady Workstation, Windows based driver for 3rd party software use
- Supported OS:
- Windows® 7, 8, 8.1, 10, Server 2008, 2008 R2, 2012, 2012 R2, 2016
- Windows® 7, 8, 8.1, 10, Server 2008, 2008 R2, 2012, 2012 R2, 2016
- Windows® 7, 8, 8.1, 10, Server 2008, 2008 R2, 2012, 2012 R2, 2016
- Windows® 7, 8, 8.1, 10, Server 2008, 2008 R2, 2012, 2012 R2, 2016
- Temperature range:
- 5...40 °C (Operating )
- 5...40 °C (Operating )
- 5...40 °C (Operating )
- 5...40 °C (Operating )