Supelco® PESTANAL® Organic Reference Standard, Atrazine-Desisopropyl
36628-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1007-28-9
- Description:Atrazine-desisopropyl
- Storage conditions:2...30 °C
- Hazard class description:H302 + H332 - H319
- Shipping conditions:STD
- MDL No.:MFCD00078645
- M-Clarity quality level:MQ100
- Synonym(s):2-Amino-4-chloro-6-ethylamino-1,3,5-triazine
- Molecular weight:173,60
- Flash point:Not applicable
- GHS signal: GHS07
: P261 - P264 - P271 - P301 + P312 - P304 + P340 + P312 - P305 + P351 + P338