ContactUsLinkComponent
Chemical resistant boot covers, AlphaTec® 2000, Model 406
Chemical resistant boot covers, AlphaTec® 2000, Model 406
Catalog # ANSEWH20B-00406-00
Supplier:  Ansell
Chemical resistant boot covers, AlphaTec® 2000, Model 406
Catalog # ANSEWH20B-00406-00
Supplier:  Ansell
Specifications
  • Shoe size:
    42 - 46
  • Garment application:
    Chemical protection
  • Colour:
    White
  • Pk:
    200 PAIR
Specifications
About this item
Superior breathable microporous laminate technology.
  • Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
  • Comfortable moisture vapor permeable fabric helps reduce the risk of heat stress
  • Optimised fit, improving wearer comfort and safety
  • Low Linting
Boot covers feature tie fastening, elastic to top of boot, and bound seams. Covers are manufactured from non-woven microporous polyethylene laminate material.
Recommended for cleanroom preparation and cleaning, production line support and maintenance, plant and machine maintenance, and veterinary services.
Certifications: Category I