ContactUsLinkComponent
Apron, sleeveless, AlphaTec® 4000, Model 212
Apron, sleeveless, AlphaTec® 4000, Model 212
Catalog # ANSEGR40W-00212-00
Supplier:  Ansell
undefined
Apron, sleeveless, AlphaTec® 4000, Model 212
Catalog # ANSEGR40W-00212-00
Supplier:  Ansell
Specifications
  • Clothing Size:
    One size
  • Colour:
    Green
  • Pk:
    100
Specifications
About this item
Innovative multi-layer chemical barrier technology.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Silicone-free
Aprons feature tie fastening to the waist with a 1000 mm long tie fastening.
Recommended for contaminated environments, chemical handling, filling/blending/charging raw materials, transferring liquids and solids, opening and draining pumps valves or lines, loading and unloading of proces equipment, raw material sample collection, concentration mixing and palletisation, Hazmat response, and large pipe assembly/disassembly.
Certifications: Type PB [3]-B; Type PB [4]-B; EN 14126; Category III
Packaging: Packaged 100 pieces/carton.