Order Entry
Belgium
ContactUsLinkComponent
Dimethyl sulphoxide, SupraSolv® for headspace gas chromatography, Supelco®
Dimethyl sulphoxide, SupraSolv® for headspace gas chromatography, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
undefined
Dimethyl sulphoxide, SupraSolv® for headspace gas chromatography, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Solubility:
    1000 g/L (20 °C)
  • Molar Mass:
    18.4 °C
  • Bulk Density:
    78.1 g/mol
  • Boiling Point:
    189 °C (1013 hPa)
  • Vapor Pressure:
    0.6 hPa (20 °C)
  • Flash Point:
    87 °C
  • Refractive Index:
    1.48 (20 °C)
  • Explosion Limit:
    1.8 - 63.0%(V)
  • Density:
    1.10 g/cm³ (20 °C)
  • Ignition Point:
    301 °C
  • Saturation Concentration:
    8.0 g/m³ (20 °C)
  • Viscosity Kinematic:
    2.14 mm²/s

Specification Test Results

Frequently Bought Together