Order Entry
Belgium
ContactUsLinkComponent
Ferrocene for synthesis, Sigma-Aldrich®
Ferrocene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Ferrocene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    172 - 175 °C
  • Molar Mass:
    186.03 g/mol
  • Bulk Density:
    500 kg/m3
  • Boiling Point:
    249 °C
  • Vapor Pressure:
    3.4 hPa (100 °C)
  • Density:
    1.49 g/cm3 (20 °C)
  • Ignition Point:
    150 °C

Specification Test Results

Related Information

Frequently Bought Together