


Ferrocene 98%
Leverancier: Thermo Fisher Scientific
Certificaten
Over dit artikel
CAS-nummer:102-54-5
:
Formule:C₁₀H₁₀Fe
MDL-nummer:MFCD00001427
:186.04 g/mol
Merck index:13,04070
Kokend Pt:249 °C (760 mmHg)
Smelten Pt:173…176 °C
Dichtheid:1,49 g/cm³ (20 °C)
Temperatuur bij opslag:Ambient
EINECS:203-039-3
UN:1325
ADR:4.1,II
:GHS02
:GHS07
:GHS09
Specificatie testresultaten
- Appearance:Orange-brown Crystalline powder or crystals (May become lumpy on storage)
- Infrared spectrum:Conforms
- Melting point:173 to 176 °C
- GC:≥97.5 %
Specificatie testresultaten
Frequently Bought Together