Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bismuth(III) nitrate basic (≥79% Bi₂O₃)

Bismuth(III) nitrate basic (≥79% Bi₂O₃)

Oxidizing
Irritant
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Certificates
Bismuth(III) nitrate basic (≥79% Bi₂O₃)
Bismuth(III) nitrate basic (≥79% Bi₂O₃)

3 Options Available Below

Product Details & Documents

CAS No.: 1304-85-4 Molecular Formula: Bi5O(OH)9(NO3)4 (approximate) Formula Weight: 1461.98 Physical Form: Powder Density: 6.04 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 260° dec. Solubility: Practically insoluble in water and alcohol. Soluble in dilute HNO3 and HCl Merck Reference: 14,1284 UN No.: 1477 MDL No.: MFCD00064844

Ceramic glazes and enamel fluxes

Synonyms:  Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
  • CAS Number:1304-85-4
  • Formula:Bi₅O(OH)₉(NO₃)₄
  • MDL Number:MFCD00064844
  • Molecular Weight:1461.99 g/mol
  • Merck Index:14,1284
  • Melting Pt:260 °C (dec.)
  • Density:6.04
  • Storage Temperature:Ambient
  • EINECS:215-136-8
  • UN:1477
  • ADR:5.1,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 46,40 €
    46,40 €
    1 * 25 g
       46,40 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 82,80 €
    82,80 €
    1 * 100 g
       82,80 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 277,00 €
    277,00 €
    1 * 500 g
       277,00 €