

Lead(II) acetate basic, anhydrous, EMSURE® for the analysis of sugar acc. to horne, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:51404-69-4
Formula:(CH₃COO)₂Pb·2Pb(OH)₂
MDL Number:MFCD00011154
Melting Pt:198…205 °C
Density:3.58 g/cm3 (22 °C)
ADR:6.1,III
:M-Clarity™ quality level = MQ300
:807.72 g/mol
Storage Temperature:Ambient
EINECS:215-630-3
UN:2291
:GHS08
:GHS09
Specification Test Results
- Assay (PbO):≥ 33.0 %
- In water insoluble matter:≤ 1.0 %
- Substances insoluble in dilute acetic acid:≤ 0.02 %
- Chloride (Cl):≤ 0.003 %
- Nitrate und Nitrite (as NO₃):passes test
- Ca (Calcium):≤ 0.01 %
- Cu (Copper):≤ 0.002 %
- Fe (Iron):≤ 0.002 %
- K (Potassium):≤ 0.02 %
- Na (Sodium):≤ 0.05 %
- Loss on Drying (105 °C):≤ 1.0 %
Specification Test Results
Frequently Bought Together