Order Entry
Belgium
ContactUsLinkComponent
Magnesium oxide, rectangular tray, Length: 100mm, Width: 50mm, Height: 12.7mm
Magnesium oxide, rectangular tray, Length: 100mm, Width: 50mm, Height: 12.7mm
Catalog # 45827.KS
CAS Number:  
Magnesium oxide, rectangular tray, Length: 100mm, Width: 50mm, Height: 12.7mm
Catalog # 45827.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :40.3 g/mol
Merck Index:13,05700
Boiling Pt:3600 °C (1013 hPa)
Melting Pt:2800 °C
Density:3.58 g/cm³ (25 °C)
EINECS:215-171-9
CAS Number:1309-48-4
Formula:MgO
MDL Number:MFCD00011109