Calcium hypochlorite
Plastic bottle for solids, 500 g
Catalog # SERA15591.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
- CAS Number:7778-54-3
- Formula:CaCl₂O₂
- MDL Number:MFCD00010900
- Molecular Weight:142.98 g/mol
- Merck Index:13,02111
- Melting Pt:∼100 °C
- Density:2.35 g/cm³ (20 °C)
- EINECS:231-908-7
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



