Order Entry
Belgium
ContactUsLinkComponent
 
 
Potassium carbonate, anhydrous ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
 
Potassium carbonate, anhydrous ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.01%
  • Assay (K₂CO₃) (by acid-base titration):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.01 %
  • Calcium (Ca):
    ≤0.002 %
  • Chloride (Cl):
    ≤0.003 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Silica (SiO₂):
    ≤0.005 %
  • Sulfur Compounds (as SO₄):
    ≤0.004 %
  • Sodium (Na):
    ≤0.02 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

 

 

Frequently Bought Together