
4-Aminoantipyrine
Catalog # SERA12373.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Merck Index:13,00596
Melting Pt:107…109 °C
:203.24 g/mol
EINECS:201-452-3
:GHS07
Storage Temperature:Ambient
CAS Number:83-07-8
Formula:C₁₁H₁₃N₃O
MDL Number:MFCD00003145