ContactUsLinkComponent
Metallographic trinocular microscopes, Panthera TEC MAT
Metallographic trinocular microscopes, Panthera TEC MAT
Supplier:  MOTIC
Certificates
undefined
Metallographic trinocular microscopes, Panthera TEC MAT
Supplier:  MOTIC
Specifications
  • Catalog No:
  • 630-3115
  • 630-3117
  • 630-3114
  • 630-3116
  • Model:
  • Panthera TEC MAT, bright field only-transmitted
  • Panthera TEC MAT, bright field and dark field-transmitted
  • Panthera TEC MAT, bright field only
  • Panthera TEC MAT, bright field and dark field
  • Observation method:
  • Brightfield, simple polarisation
  • Brightfield, darkfield and simple polarisation
  • Brightfield, simple polarisation
  • Brightfield, darkfield and simple polarisation
  • Head:
  • 25° inclined, 360° rotating (light split 50:50 fixed)
  • 25° inclined, 360° rotating (light split 50:50 fixed)
  • 25° inclined, 360° rotating (light split 50:50 fixed)
  • 25° inclined, 360° rotating (light split 50:50 fixed)
  • Eyepieces:
  • UC-WF 10×/22 mm
  • UC-WF 10×/22 mm
  • UC-WF 10×/22 mm
  • UC-WF 10×/22 mm
  • Nosepiece:
  • Quintuple, reversed
  • Quintuple, reversed
  • Quintuple, reversed
  • Quintuple, reversed
  • Objectives:
  • Plan achromatic BD 5×, 10×, 20×, Plan S-APO BD 50×
  • Plan achromatic BD 5×, 10×, 20×, Plan S-APO BD 50×
  • Plan achromatic BD 5×, 10×, 20×, Plan S-APO BD 50×
  • Plan achromatic BD 5×, 10×, 20×, Plan S-APO BD 50×
  • Stage:
  • Mechanical stage 300×180 mm surface, hard coated, glass plate (travel range 150×100 mm)
  • Mechanical stage 300×180 mm surface, hard coated, glass plate (travel range 150×100 mm)
  • Mechanical stage 300×180 mm surface, hard coated, glass plate (travel range 150×100 mm)
  • Mechanical stage 300×180 mm surface, hard coated, glass plate (travel range 150×100 mm)
  • Focusing system:
  • Coaxial coarse and fine focusing system with tension adjustment
  • Coaxial coarse and fine focusing system with tension adjustment
  • Coaxial coarse and fine focusing system with tension adjustment
  • Coaxial coarse and fine focusing system with tension adjustment
Specifications
Eyepieces
Product Information
Frequently Bought Together