1-(3-Chlorophenyl)piperazine
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:6640-24-0
:
Formula:C₁₀H₁₃ClN₂
MDL Number:MFCD00040732
Boiling Pt:336 °C (1013 hPa)
Melting Pt:210 °C
Density:1.1897 g/cm³ (25 °C)
:196.68 g/mol
EINECS:229-654-7
Frequently Bought Together