PIPES ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 5625-37-6 Molecular Formula: C8H18N2O6S2 Formula Weight: 302.37 Melting Point: 300° Beilstein Reference: 817713 Merck Reference: 14,7479 MDL No.: MFCD00006159
Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
- CAS Number:5625-37-6
- Formula:C₈H₁₈N₂O₆S₂
- MDL Number:MFCD00006159
- Molecular Weight:302.37 g/mol
- Merck Index:14,7479
- Boiling Pt:499 °C (1013 hPa)
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:227-057-6
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 5625-37-6 Molecular Formula: C8H18N2O6S2 Formula Weight: 302.37 Melting Point: 300° Beilstein Reference: 817713 Merck Reference: 14,7479 MDL No.: MFCD00006159
Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
- CAS Number:5625-37-6
- Formula:C₈H₁₈N₂O₆S₂
- MDL Number:MFCD00006159
- Molecular Weight:302.37 g/mol
- Merck Index:14,7479
- Boiling Pt:499 °C (1013 hPa)
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:227-057-6
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



